Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108863.1 | NP_001102333.1 | 1, 2 | 383 | ||
XM_006251518.2 | XP_006251580.1 | 3, 4 | 515 | |||
XM_008770433.1 | XP_008768655.1 | 2, 3 | 1691 | |||
XM_008770434.1 | XP_008768656.1 | 3, 4 | 1053 |