Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acyp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169145.1 | NP_001162616.1 | 3 | 334 | ||
XM_008770465.1 | XP_008768687.1 | 3 | 432 | |||
XM_008770466.1 | XP_008768688.1 | 3 | 415 | |||
GenBank mRNA | CB613181.1 | - | 3 | 344 | ||
CO404339.1 | - | 3 | 165 | |||
FQ215015.1 | - | 3 | 166 | |||
FQ224926.1 | - | 3 | 202 |