Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108865.1 | NP_001102335.1 | 2 | 394 | ||
GenBank mRNA | AB002591.1 | - | 2 | 645 | ||
FQ215794.1 | - | 2 | 388 | |||
FQ229094.1 | - | 2 | 376 |