Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008561.1 | NP_001008561.1 | 2 | 582 | ||
XM_006251890.2 | XP_006251952.1 | 2 | 1059 | |||
XM_006251891.2 | XP_006251953.1 | 2 | 981 | |||
GenBank mRNA | AY303815.1 | - | 2 | 582 |