Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029904.1 | NP_001025075.1 | 619 | |||
GenBank mRNA | J00771.1 | - | 2 | 613 |
Gene Symbol: LOC103690354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770630.1 | XP_008768852.1 | 2 | 628 |
Gene Symbol: LOC103694853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773795.1 | XP_008772017.1 | 658 |
Gene Symbol: Rnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251893.2 | XP_006251955.1 | 2 | 675 |
Gene Symbol: Rnase1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251875.1 | XP_006251937.1 | 2 | 658 |