Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1311300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF196199.1 | - | 4 | 704 | ||
AF196210.1 | - | 4 | 602 | |||
AF196211.1 | - | 4 | 631 | |||
AF196212.1 | - | 4 | 602 | |||
AF196213.1 | - | 3 | 594 | |||
AF196214.1 | - | 3 | 532 | |||
AF196215.1 | - | 3 | 605 | |||
AF196216.1 | - | 3 | 605 | |||
AF196217.1 | - | 641 | ||||
AF196218.1 | - | 648 | ||||
AF196219.1 | - | 641 | ||||
AF196220.1 | - | 641 | ||||
AF196221.1 | - | 645 | ||||
AF196222.1 | - | 644 | ||||
AF196223.1 | - | 560 | ||||
AF196224.1 | - | 659 | ||||
AF196225.1 | - | 673 | ||||
AF196226.1 | - | 681 | ||||
AF196227.1 | - | 535 | ||||
AF196228.1 | - | 645 | ||||
AF196229.1 | - | 5 | 577 | |||
AF196230.1 | - | 3 | 362 | |||
AF196231.1 | - | 4 | 673 | |||
AF196232.1 | - | 6 | 576 | |||
AF196233.1 | - | 4 | 594 | |||
AF196234.1 | - | 4 | 658 | |||
AF196235.1 | - | 4 | 484 | |||
AF196236.1 | - | 4 | 684 | |||
AF196237.1 | - | 5 | 563 | |||
AF196238.1 | - | 6 | 587 | |||
AF196239.1 | - | 5 | 705 | |||
AF196240.1 | - | 5 | 591 | |||
AF196242.1 | - | 4 | 669 | |||
AF196243.1 | - | 5 | 565 | |||
AF196244.1 | - | 6 | 554 | |||
AF196245.1 | - | 5 | 643 | |||
AF196246.1 | - | 3 | 613 | |||
AF196247.1 | - | 6 | 554 | |||
AF196266.1 | - | 2 | 263 | |||
AJ249228.1 | - | 4 | 349 | |||
BC099216.1 | - | 4 | 704 | 7384680 |