Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108866.1 | NP_001102336.1 | 3 | 219 | ||
XM_006251995.2 | XP_006252057.1 | 3 | 424 | |||
XM_006251996.2 | XP_006252058.1 | 3 | 215 |