Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108867.2 | NP_001102337.2 | 3 | 261 | ||
GenBank mRNA | CB814013.1 | - | 3 | 272 | ||
CO573950.1 | - | 1 | 61 |