Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014235.1 | NP_001014257.1 | 411 | |||
GenBank mRNA | BC088253.1 | - | 411 | 7374374 |