Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006308.2 | NP_006299.1 | 1 | 519 | ||
GenBank mRNA | AK314215.1 | - | 1 | 520 | ||
BC126272.1 | - | 1 | 374 | 8991988 | ||
BC132869.1 | - | 1 | 386 | 40146891 | ||
BX106905.1 | - | 1 | 504 | |||
CR450314.1 | - | 1 | 330 | |||
U15590.1 | - | 1 | 505 | |||
Y17782.1 | - | 1 | 478 |