Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scara3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108870.1 | NP_001102340.1 | 5 | 1248 | ||
GenBank mRNA | FQ211593.1 | - | 5 | 1050 |