Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dzip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252465.2 | XP_006252527.1 | 15 | 1891 | ||
XM_006252466.1 | XP_006252528.1 | 15 | 1764 | |||
XM_008770948.1 | XP_008769170.1 | 14 | 1944 | |||
XM_008770949.1 | XP_008769171.1 | 14 | 1941 | |||
GenBank mRNA | BC089964.1 | - | 2 | 82 |