Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258288.1 | NP_001245217.1 | 5 | 532 | ||
NM_001258289.1 | NP_001245218.1 | 6 | 927 | |||
NM_003944.3 | NP_003935.2 | 6 | 718 | |||
GenBank mRNA | AK023875.1 | - | 4 | 1100 | ||
AK296661.1 | - | 6 | 896 | |||
AK298355.1 | - | 6 | 583 | |||
AK303815.1 | - | 5 | 498 | |||
AK315643.1 | - | 6 | 688 | |||
BC009084.1 | - | 6 | 672 | 3853674 | ||
BC032997.1 | - | 4 | 1103 | 4839079 | ||
CR456852.1 | - | 5 | 625 | |||
EU794660.1 | - | 6 | 672 | |||
U29091.1 | - | 5 | 629 |