Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258289.1 | NP_001245218.1 | 4 | 517 | ||
NM_003944.3 | NP_003935.2 | 4 | 308 | |||
XM_017002757.1 | XP_016858246.1 | 4 | 510 | |||
GenBank mRNA | AK023875.1 | - | 2 | 690 | ||
AK296661.1 | - | 4 | 486 | |||
AK298355.1 | - | 4 | 279 | |||
AK309264.1 | - | 2 | 851 | |||
AK310044.1 | - | 3 | 717 | |||
AK315643.1 | - | 4 | 278 | |||
BC009084.1 | - | 4 | 262 | 3853674 | ||
BC032997.1 | - | 2 | 693 | 4839079 | ||
CR456852.1 | - | 3 | 215 | |||
EU794660.1 | - | 4 | 262 | |||
U29091.1 | - | 3 | 219 |