Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014236.1 | NP_001014258.1 | 2, 3 | 273 | ||
XM_006252912.2 | XP_006252974.1 | 2, 3 | 286 | |||
XM_008771139.1 | XP_008769361.1 | 2, 3 | 288 | |||
XM_008771140.1 | XP_008769362.1 | 2, 3 | 286 | |||
XM_008771141.1 | XP_008769363.1 | 2, 3 | 286 | |||
XM_008771142.1 | XP_008769364.1 | 2, 3 | 286 | |||
GenBank mRNA | BC079147.1 | - | 2, 3 | 273 | 7128392 |