Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258288.1 | NP_001245217.1 | 3 | 166 | ||
NM_001258289.1 | NP_001245218.1 | 3 | 375 | |||
NM_003944.3 | NP_003935.2 | 3 | 166 | |||
XM_017002757.1 | XP_016858246.1 | 3 | 368 | |||
GenBank mRNA | AK023875.1 | - | 2 | 295 | ||
AK296661.1 | - | 3 | 344 | |||
AK298355.1 | - | 3 | 137 | |||
AK303815.1 | - | 3 | 132 | |||
AK309264.1 | - | 2 | 457 | |||
AK310044.1 | - | 3 | 322 | |||
AK315643.1 | - | 3 | 136 | |||
BC009084.1 | - | 3 | 120 | 3853674 | ||
BC032997.1 | - | 2 | 298 | 4839079 | ||
CR456852.1 | - | 2 | 73 | |||
DB288439.1 | - | 2 | 324 | |||
EU794660.1 | - | 3 | 120 | |||
U29091.1 | - | 2 | 77 |