Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004045.3 | NP_004036.1 | 2, 3 | 189 | ||
GenBank mRNA | AK128210.1 | - | 2, 3 | 1066 | ||
AK293063.1 | - | 2, 3 | 196 | |||
BC112248.1 | - | 2, 3 | 271 | 8327716 | ||
BC112250.1 | - | 2, 3 | 271 | 8327718 | ||
BI599233.1 | - | 2, 3 | 197 | |||
BT009786.1 | - | 1, 2 | 91 | |||
BU172675.1 | - | 2, 3 | 567 | |||
CN280769.1 | - | 2, 3 | 166 | |||
CR456901.1 | - | 1, 2 | 91 |