Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033710.1 | NP_001028882.1 | 2, 3 | 255 | ||
GenBank mRNA | BC104722.1 | - | 2, 3 | 255 | 7383387 |