Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003948.3 | NP_003939.1 | 4 | 1052 | ||
XM_005263335.2 | XP_005263392.1 | 4 | 771 | |||
XM_006714406.2 | XP_006714469.1 | 4 | 771 | |||
XM_017008809.1 | XP_016864298.1 | 4 | 771 | |||
XM_017008810.1 | XP_016864299.1 | 4 | 771 | |||
XM_017008811.1 | XP_016864300.1 | 4 | 771 | |||
GenBank mRNA | AK294866.1 | - | 4 | 1038 | ||
AK312490.1 | - | 4 | 1043 | |||
BC093646.1 | - | 4 | 576 | 7939491 | ||
BC093981.1 | - | 4 | 576 | 7939826 | ||
U35146.1 | - | 4 | 902 |