Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039036.1 | NP_001034125.1 | 4, 5 | 618 | ||
XM_008762189.1 | XP_008760411.1 | 4, 5 | 1161 | |||
XM_008762190.1 | XP_008760412.1 | 3, 4 | 3164 | |||
GenBank mRNA | BC098859.1 | - | 4, 5 | 618 | 7380252 |
Gene Symbol: Gabpb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135015.1 | NP_001128487.1 | 1 | 469 | ||
XM_006254075.2 | XP_006254137.1 | 2 | 502 |