Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108886.1 | NP_001102356.1 | 2 | 247 | ||
GenBank mRNA | FQ228021.1 | - | 2 | 218 |