Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F2RL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003950.2 | NP_003941.2 | 2 | 1178 | ||
XM_005260139.2 | XP_005260196.1 | 2 | 999 | |||
GenBank mRNA | AF055917.1 | - | 2 | 1178 | ||
AF080214.1 | - | 2 | 1005 | |||
AY431102.1 | - | 2 | 1003 | |||
BC074782.2 | - | 2 | 1022 | 30915337 |