Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108888.2 | NP_001102358.2 | 2, 3 | 303 | ||
GenBank mRNA | BC167013.1 | - | 2, 3 | 228 | 9032688 | |
BE126660.1 | - | 2, 3 | 303 | |||
FQ213789.1 | - | 2, 3 | 235 | |||
FQ235123.1 | - | 2, 3 | 235 |