Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282195.1 | NP_001269124.1 | 6, 7 | 751 | ||
NM_001282196.1 | NP_001269125.1 | 6, 7 | 742 | |||
NM_001282197.1 | NP_001269126.1 | 6, 7 | 742 | |||
NM_001282198.1 | NP_001269127.1 | 5, 6 | 504 | |||
XM_005262489.4 | XP_005262546.1 | 3, 4 | 253 | |||
XM_011531402.2 | XP_011529704.1 | 6, 7 | 753 | |||
XM_011531403.1 | XP_011529705.1 | 5, 6 | 467 | |||
XM_017029936.1 | XP_016885425.1 | 5, 6 | 467 | |||
XM_017029937.1 | XP_016885426.1 | 6, 7 | 753 | |||
XM_017029938.1 | XP_016885427.1 | 6, 7 | 748 | |||
XM_017029939.1 | XP_016885428.1 | 6, 7 | 717 | |||
XM_017029940.1 | XP_016885429.1 | 3, 4 | 258 | |||
XM_017029941.1 | XP_016885430.1 | 7, 8 | 770 | |||
XM_017029942.1 | XP_016885431.1 | 3, 4 | 263 | |||
GenBank mRNA | AF078544.1 | - | 5, 6 | 747 | ||
AF155809.1 | - | 5, 6 | 490 | |||
AF155810.1 | - | 5, 6 | 481 | |||
AF155811.1 | - | 5, 6 | 481 | |||
AK296155.1 | - | 5, 6 | 434 | |||
AK297499.1 | - | 6, 7 | 685 | |||
AK309433.1 | - | 6, 7 | 676 | |||
AY358099.1 | - | 5, 6 | 499 | |||
BC119666.2 | - | 5, 6 | 606 | 40116117 | ||
BC119667.1 | - | 5, 6 | 606 | 40116118 |