Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000538.1 | NP_001000538.1 | 1 | 784 |
Gene Symbol: LOC103689958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758874.1 | XP_008757096.1 | 1 | 784 |