Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100912013 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748754.2 | XP_003748802.1 | 2 | 1074 |
Gene Symbol: Nlrp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_344856.5 | XP_344857.4 | 1074 |