Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108906.1 | NP_001102376.1 | 2 | 222 | ||
XM_006228117.1 | XP_006228179.1 | 2 | 612 | |||
XM_006228118.1 | XP_006228180.1 | 2 | 612 | |||
GenBank mRNA | BC167093.1 | - | 2 | 220 | 9088608 | |
FQ222002.1 | - | 2 | 188 | |||
FQ227339.1 | - | 2 | 238 | |||
FQ229914.1 | - | 2 | 130 | |||
FQ230074.1 | - | 2 | 223 | |||
FQ230732.1 | - | 2 | 238 | |||
FQ231086.1 | - | 2 | 243 | |||
FQ231147.1 | - | 2 | 217 | |||
FQ233974.1 | - | 2 | 133 |