Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000701.7 | NP_000692.2 | 6 | 860 | ||
NM_001160233.1 | NP_001153705.1 | 6 | 884 | |||
NM_001160234.1 | NP_001153706.1 | 6 | 693 | |||
XM_017001360.1 | XP_016856849.1 | 7 | 4079 | |||
XM_017001361.1 | XP_016856850.1 | 7 | 3421 | |||
GenBank mRNA | AK293689.1 | - | 3 | 265 | ||
AK295095.1 | - | 6 | 883 | |||
AK296362.1 | - | 6 | 693 | |||
AK296384.1 | - | 6 | 819 | |||
AK308418.1 | - | 6 | 625 | |||
AK314777.1 | - | 6 | 705 | |||
AL531892.3 | - | 6 | 827 | |||
BC003077.2 | - | 6 | 773 | 3507685 | ||
BC050359.1 | - | 6 | 771 | 6065209 | ||
U16798.1 | - | 5 | 521 |