Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108907.1 | NP_001102377.1 | 4 | 503 | ||
XM_006228328.2 | XP_006228390.1 | 4 | 521 | |||
XM_006228329.2 | XP_006228391.1 | 4 | 821 | |||
XM_006228330.2 | XP_006228392.1 | 4 | 726 | |||
XM_006228331.2 | XP_006228393.1 | 4 | 1593 | |||
XM_006228332.2 | XP_006228394.1 | 4 | 1107 | |||
XM_006228333.2 | XP_006228395.1 | 3 | 1562 |