Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004669.2 | NP_004660.2 | 4 | 369 | ||
XM_017015281.1 | XP_016870770.1 | 4 | 574 | |||
XM_017015282.1 | XP_016870771.1 | 4 | 537 | |||
GenBank mRNA | AF102166.1 | - | 4 | 329 | ||
BC007012.1 | - | 4 | 365 | 3996920 | ||
CR542066.1 | - | 3 | 253 |