Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sergef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229227.2 | XP_006229289.1 | 8 | 812 | ||
XM_006229228.2 | XP_006229290.1 | 7 | 749 | |||
XM_344891.7 | XP_344892.4 | 8 | 812 |