Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759595.1 | XP_008757817.1 | 1 | 1737 |
Gene Symbol: Bnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108916.1 | NP_001102386.1 | 4 | 1157 |