Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002027.2 | NP_002018.1 | 5, 6 | 679 | ||
NR_033698.1 | 4, 5 | 593 | ||||
GenBank mRNA | AK123002.1 | - | 5, 6 | 1066 | ||
AK292121.1 | - | 5, 6 | 655 | |||
AL698961.1 | - | 3, 4 | 362 | |||
BC017029.2 | - | 5, 6 | 622 | |||
BC084566.1 | - | 5, 6 | 639 | 6423445 | ||
BT009854.1 | - | 5, 6 | 631 | |||
L00634.1 | - | 5, 6 | 647 | |||
L10413.1 | - | 5, 6 | 637 |