Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278720.1 | NP_001265649.1 | 3, 4 | 543 | ||
NM_001318733.1 | NP_001305662.1 | 2, 3 | 640 | |||
XM_017023849.1 | XP_016879338.1 | 2, 3 | 643 | |||
XM_017023850.1 | XP_016879339.1 | 2, 3 | 643 | |||
GenBank mRNA | AJ272344.1 | - | 3, 4 | 503 | ||
AK294136.1 | - | 3, 4 | 487 | |||
BC120874.2 | - | 2, 3 | 632 | 40081041 | ||
BC120875.2 | - | 2, 3 | 633 | 40081042 | ||
BI600093.1 | - | 3, 4 | 551 | |||
Y17108.1 | - | 2, 3 | 640 |