Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077675.1 | NP_001071143.1 | 7 | 1467 | ||
GenBank mRNA | BC127539.1 | - | 7 | 1467 | 7192024 | |
FQ224850.1 | - | 7 | 1509 |