Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000702.3 | NP_000693.1 | 6, 7 | 771 | ||
GenBank mRNA | AB018321.2 | - | 6, 7 | 728 | ||
AK091617.1 | - | 6, 7 | 742 | |||
AK124581.1 | - | 4, 5 | 443 | |||
AK295048.1 | - | 3, 4 | 280 | |||
AK314296.1 | - | 6, 7 | 742 | |||
AL831991.1 | - | 5, 6 | 560 | |||
AL831997.1 | - | 5, 6 | 586 | |||
BC052271.1 | - | 6, 7 | 722 | 6672133 | ||
KU177927.1 | - | 5, 6 | 639 | |||
KU177928.1 | - | 5, 6 | 639 |