Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014248.1 | NP_001014270.1 | 5 | 1002 | ||
XM_006231678.2 | XP_006231740.1 | 5 | 1162 | |||
GenBank mRNA | BC083839.1 | - | 5 | 1002 | 7114271 |