Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108943.2 | NP_001102413.1 | 4 | 353 | ||
GenBank mRNA | DQ387062.1 | - | 4 | 353 |