Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014254.1 | NP_001014276.1 | 13 | 1628 | ||
XM_008761185.1 | XP_008759407.1 | 13 | 1833 | |||
XM_008761186.1 | XP_008759408.1 | 14 | 1700 | |||
XM_008761187.1 | XP_008759409.1 | 13 | 1830 | |||
XM_008761188.1 | XP_008759410.1 | 13 | 1812 | |||
XM_008761189.1 | XP_008759411.1 | 13 | 1809 | |||
XM_008761190.1 | XP_008759412.1 | 13 | 1809 | |||
XM_008761191.1 | XP_008759413.1 | 13 | 1806 | |||
XM_008761192.1 | XP_008759414.1 | 13 | 1679 | |||
XM_008761193.1 | XP_008759415.1 | 10 | 1480 | |||
XM_008761194.1 | XP_008759416.1 | 10 | 959 | |||
GenBank mRNA | BC083788.1 | - | 13 | 1628 | 7098251 |