Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100986.1 | NP_001094456.1 | 2 | 378 | ||
XM_006233090.2 | XP_006233152.1 | 2 | 329 | |||
XM_006233097.2 | XP_006233159.1 | 2 | 328 | |||
XM_008761412.1 | XP_008759634.1 | 2 | 339 | |||
XM_008761413.1 | XP_008759635.1 | 2 | 1038 | |||
XM_008761414.1 | XP_008759636.1 | 3 | 1193 | |||
XM_008761415.1 | XP_008759637.1 | 3 | 1524 | |||
XM_008761416.1 | XP_008759638.1 | 3 | 863 | |||
XM_008761417.1 | XP_008759639.1 | 2 | 541 | |||
XM_008761418.1 | XP_008759640.1 | 2 | 643 | |||
XM_008761419.1 | XP_008759641.1 | 2 | 340 | |||
XM_008761420.1 | XP_008759642.1 | 2 | 1033 | |||
GenBank mRNA | EF460311.1 | - | 2 | 378 | ||
EF460312.1 | - | 2 | 378 |