Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317800.1 | NP_001304729.1 | 2 | 193 | ||
NM_003974.3 | NP_003965.2 | 4 | 563 | |||
NM_201349.2 | NP_958728.1 | 3 | 475 | |||
XM_005273680.4 | XP_005273737.1 | 3 | 290 | |||
GenBank mRNA | AK226110.1 | - | 2 | 163 | ||
AK292126.1 | - | 4 | 519 | |||
AK296671.1 | - | 3 | 457 | |||
BC032623.1 | - | 4 | 531 | 5242017 | ||
BQ067133.1 | - | 489 | ||||
CB270269.1 | - | 2 | 147 | |||
DC295259.1 | - | 3 | 475 |