Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161441.1 | NP_001154913.1 | 7 | 1021 | ||
NM_001161442.1 | NP_001154914.1 | 7 | 960 | |||
NM_001161443.1 | NP_001154915.1 | 6 | 870 | |||
NM_001161444.1 | NP_001154916.1 | 7 | 991 | |||
NM_003975.3 | NP_003966.2 | 7 | 991 | |||
XM_006711615.2 | XP_006711678.1 | 7 | 1009 | |||
XM_017002762.1 | XP_016858251.1 | 7 | 1038 | |||
XM_017002763.1 | XP_016858252.1 | 7 | 1029 | |||
XM_017002764.1 | XP_016858253.1 | 7 | 976 | |||
XM_017002765.1 | XP_016858254.1 | 7 | 989 | |||
XM_017002766.1 | XP_016858255.1 | 7 | 979 | |||
GenBank mRNA | AF051325.1 | - | 7 | 913 | ||
AF097744.1 | - | 7 | 890 | |||
AJ000553.1 | - | 7 | 937 | |||
AK222737.1 | - | 7 | 914 | |||
AK313706.1 | - | 7 | 985 | |||
AY763098.1 | - | 6 | 870 | |||
AY763099.1 | - | 7 | 967 | |||
AY763100.1 | - | 7 | 897 | |||
BC012107.1 | - | 7 | 953 | 4636736 |