Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128183.1 | NP_001121655.1 | 2 | 221 | ||
XM_006233632.2 | XP_006233694.2 | 2 | 362 | |||
GenBank mRNA | DQ537491.1 | - | 2 | 207 |