Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128183.1 | NP_001121655.1 | 3 | 258 | ||
XM_006233632.2 | XP_006233694.2 | 3 | 399 | |||
GenBank mRNA | DQ537491.1 | - | 3 | 244 |