Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136215.1 | NP_001129687.1 | 4 | 1710 | ||
NM_057090.2 | NP_476431.2 | 5 | 1859 | |||
NM_057091.2 | NP_476432.2 | 5 | 1835 | |||
GenBank mRNA | AF120274.1 | - | 5 | 1835 | ||
BC062375.1 | - | 4 | 1218 | 5453642 |