Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233905.2 | XP_006233967.1 | 24 | 2540 | ||
XM_006233906.2 | XP_006233968.1 | 24 | 3030 | |||
XM_345335.7 | XP_345336.2 | 24 | 2540 |