Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSTPIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024430.3 | NP_077748.3 | 2 | 158 | ||
XM_011526253.2 | XP_011524555.1 | 2 | 201 | |||
XM_011526254.1 | XP_011524556.1 | 2 | 134 | |||
GenBank mRNA | BC035395.1 | - | 2 | 131 | 4524856 | |
BP352927.1 | - | 2 | 158 | |||
KU178455.1 | - | 2 | 62 | |||
KU178456.1 | - | 2 | 62 | |||
KU178457.1 | - | 2 | 62 |