Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000563.1 | NP_001000563.1 | 1 | 899 | ||
XM_008761992.1 | XP_008760214.1 | 2 | 943 |
Gene Symbol: Olr526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001375.1 | NP_001001375.1 | 1 | 899 |
Gene Symbol: Olr530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001376.1 | NP_001001376.1 | 1 | 899 |