Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014258.1 | NP_001014280.1 | 2, 3 | 578 | ||
XM_006235078.2 | XP_006235140.1 | 2, 3 | 772 | |||
XM_006235079.2 | XP_006235141.1 | 2, 3 | 761 | |||
XM_006235080.2 | XP_006235142.1 | 2, 3 | 261 | |||
GenBank mRNA | BC085849.1 | - | 2, 3 | 578 | 7189638 |