Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198608.1 | NP_001185537.1 | 6 | 872 | ||
NM_001198609.1 | NP_001185538.1 | 6 | 872 | |||
NM_001198614.1 | NP_001185543.1 | 7 | 875 | |||
NM_001198615.1 | NP_001185544.1 | 6 | 605 | |||
NM_001198617.1 | NP_001185546.1 | 4 | 686 | |||
NM_001198618.1 | NP_001185547.1 | 6 | 735 | |||
NM_001198619.1 | NP_001185548.1 | 6 | 608 | |||
NM_003980.4 | NP_003971.1 | 6 | 968 | |||
XM_006715598.2 | XP_006715661.1 | 7 | 1393 | |||
XM_006715599.2 | XP_006715662.1 | 7 | 1162 | |||
XM_006715600.2 | XP_006715663.1 | 7 | 1162 | |||
XM_006715601.3 | XP_006715664.1 | 6 | 1164 | |||
XM_011536243.2 | XP_011534545.1 | 6 | 1763 | |||
GenBank mRNA | AJ242501.1 | - | 4 | 275 | ||
AK295749.1 | - | 6 | 600 | |||
AK296483.1 | - | 6 | 735 | |||
AK296556.1 | - | 6 | 734 | |||
AK299355.1 | - | 6 | 605 | |||
AK307873.1 | - | 6 | 711 | |||
AK311114.1 | - | 4 | 234 | |||
AK311234.1 | - | 7 | 752 | |||
AK311236.1 | - | 7 | 710 | |||
AK312789.1 | - | 6 | 568 | |||
AK316062.1 | - | 6 | 608 | |||
AK316529.1 | - | 7 | 741 | |||
BC025777.1 | - | 6 | 564 | 5222987 | ||
X73882.1 | - | 6 | 657 |